missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bcl-2-like protein 11 Protein
A DNA sequence encoding the Homo sapiens (Human) Bcl-2-like protein 11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5712-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001191035.1 | |
Bcl-2-like protein 11 Protein | |
Research Use Only | |
BCL2L11 | |
Recombinant Protein | |
E. coli | |
MAKQPSDVSS ECDREGRQLQ PAERPPQLRP GAPTSLQTEP QGNPEGNHGG EGDSCPHGSP QGPLAPPASP GPFATRSPLF IFMRRSSLLS RSSSGYFSFD TDRSPAPMSC DKSTQTPSPP CQAFNHYLSA MASMRQAEPA DMRPEIWIAQ ELRRIGDEFN AYYARRVFLN NYQAAEDHPR MVILRLLRYI VRLVWRMH | |
Greater than 90% as determined by SDS-PAGE. |
10018 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |