missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bcl-2-like protein 11 Protein
A DNA sequence encoding the Homo sapiens (Human) Bcl-2-like protein 11, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001191035.1 |
---|---|
Gene ID (Entrez) | 10018 |
Name | Bcl-2-like protein 11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15927199
|
enQuireBio™
QP5712-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947199
|
enQuireBio™
QP5712-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937199
|
enQuireBio™
QP5712-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15957199
|
enQuireBio™
QP5712-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15967199
|
enQuireBio™
QP5712-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15977199
|
enQuireBio™
QP5712-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001191035.1 | |
Bcl-2-like protein 11 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10018 | |
Research Use Only | |
BCL2L11 | |
Recombinant Protein | |
E. coli | |
MAKQPSDVSS ECDREGRQLQ PAERPPQLRP GAPTSLQTEP QGNPEGNHGG EGDSCPHGSP QGPLAPPASP GPFATRSPLF IFMRRSSLLS RSSSGYFSFD TDRSPAPMSC DKSTQTPSPP CQAFNHYLSA MASMRQAEPA DMRPEIWIAQ ELRRIGDEFN AYYARRVFLN NYQAAEDHPR MVILRLLRYI VRLVWRMH | |
Greater than 90% as determined by SDS-PAGE. |