missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bcl 2 (1-206 a. a. ) Protein
A cDNA sequence encoding the Bcl 2 (1-206 a. a. ) was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10490-500ug
Additional Details : Weight : 0.01000kg
Specifications
Bcl 2 (1-206 a. a. ) Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
DPBS pH 7.4, 0.2microm filter sterilised. |
500 μg | |
Untested | |
Tick-borne encephalitis virus | |
His | |
>90% as determined by SDS-PAGE. |