missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bcl 2 (1-206 a. a. ) Protein
A cDNA sequence encoding the Bcl 2 (1-206 a. a. ) was constructed and used to recombinantly synthesize the protein.
£421.00 - £1525.00
Specifications
Name | Bcl 2 (1-206 a. a. ) Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Untested |
Product Type | Recombinant Protein |
Cross Reactivity | Tick-borne encephalitis virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15964018
|
enQuireBio™
QP10490-100UG |
100 μg |
£421.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974018
|
enQuireBio™
QP10490-500UG |
500 μg |
£1525.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Bcl 2 (1-206 a. a. ) Protein | |
Untested | |
Tick-borne encephalitis virus | |
His | |
>90% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Human | |
DPBS pH 7.4, 0.2microm filter sterilised. |