missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BABAM1 Protein
A cDNA sequence encoding the BABAM1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11122-20ug
Additional Details : Weight : 0.01000kg
Specifications
BABAM1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEVAEPS SPTEEEEEEE EHSAEPRPRT RSNPEGAEDR AVGAQASVGS RSEGEGEAAS ADDGSLNTSG AGPKSWQVPP PAPEVQIRTP RVNCPEKVII CLDLSEEMSL PKLESFNGSK TNALNVSQKM IEMFVRTKHK IDKSHEFALV VVNDDTAWLS GLTSDPRELC SCLYDLETAS CSTFNLEGLF SLIQQKTELP VTENVQTIPP PYVVRTILVY SRPPCQPQFS LTEPMKKMFQ CPYFFFDVVY IHNGTEEKEE EMSWKDMFAF MGSLDTKGTS YKYEVALAGP ALELHNCMAK LLAHPLQRPC QSHASYSLLE EEDEAIEVEA TV | |
Greater than 85% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BABAM1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |