missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BABAM1 Protein
A cDNA sequence encoding the BABAM1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | BABAM1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986218
|
enQuireBio™
QP11122-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976218
|
enQuireBio™
QP11122-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966218
|
enQuireBio™
QP11122-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifica
BABAM1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BABAM1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEVAEPS SPTEEEEEEE EHSAEPRPRT RSNPEGAEDR AVGAQASVGS RSEGEGEAAS ADDGSLNTSG AGPKSWQVPP PAPEVQIRTP RVNCPEKVII CLDLSEEMSL PKLESFNGSK TNALNVSQKM IEMFVRTKHK IDKSHEFALV VVNDDTAWLS GLTSDPRELC SCLYDLETAS CSTFNLEGLF SLIQQKTELP VTENVQTIPP PYVVRTILVY SRPPCQPQFS LTEPMKKMFQ CPYFFFDVVY IHNGTEEKEE EMSWKDMFAF MGSLDTKGTS YKYEVALAGP ALELHNCMAK LLAHPLQRPC QSHASYSLLE EEDEAIEVEA TV | |
Greater than 85% as determined by SDS-PAGE. |