missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Arginase-2, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Arginase-2, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5676-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001163.1 | |
Arginase-2, mitochondrial Protein | |
Research Use Only | |
Arg2 | |
Recombinant Protein | |
E. coli | |
VHSVAVIGAP FSQGQKRKGV EHGPAAIREA GLMKRLSSLG CHLKDFGDLS FTPVPKDDLY NNLIVNPRSV GLANQELAEV VSRAVSDGYS CVTLGGDHSL AIGTISGHAR HCPDLCVVWV DAHADINTPL TTSSGNLHGQ PVSFLLRELQ DKVPQLPGFS WIKPCISSAS IVYIGLRDVD PPEHFILKNY DIQYFSMRDI DRLGIQKVME RTFDLLIGKR QRPIHLSFDI DAFDPTLAPA TGTPVVGGLT YREGMYIAEE IHNTGLLSAL DLVEVNPQLA TSEEEAKTTA NLAVDVIASS FGQTREGGHI VYDQLPTPSS PDESENQARV RI | |
Greater than 80% as determined by SDS-PAGE. |
384 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |