missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Arginase-2, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Arginase-2, mitochondrial, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001163.1 |
---|---|
Gene ID (Entrez) | 384 |
Name | Arginase-2, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966919
|
enQuireBio™
QP5676-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986919
|
enQuireBio™
QP5676-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976919
|
enQuireBio™
QP5676-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15996919
|
enQuireBio™
QP5676-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15906929
|
enQuireBio™
QP5676-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15916929
|
enQuireBio™
QP5676-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001163.1 | |
Arginase-2, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
384 | |
Research Use Only | |
Arg2 | |
Recombinant Protein | |
E. coli | |
VHSVAVIGAP FSQGQKRKGV EHGPAAIREA GLMKRLSSLG CHLKDFGDLS FTPVPKDDLY NNLIVNPRSV GLANQELAEV VSRAVSDGYS CVTLGGDHSL AIGTISGHAR HCPDLCVVWV DAHADINTPL TTSSGNLHGQ PVSFLLRELQ DKVPQLPGFS WIKPCISSAS IVYIGLRDVD PPEHFILKNY DIQYFSMRDI DRLGIQKVME RTFDLLIGKR QRPIHLSFDI DAFDPTLAPA TGTPVVGGLT YREGMYIAEE IHNTGLLSAL DLVEVNPQLA TSEEEAKTTA NLAVDVIASS FGQTREGGHI VYDQLPTPSS PDESENQARV RI | |
Greater than 80% as determined by SDS-PAGE. |