missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apolipoprotein C-II Protein
A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein C-II, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5665-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000474.2 | |
Apolipoprotein C-II Protein | |
Research Use Only | |
APOC2 | |
Recombinant Protein | |
E. coli | |
TQQPQQDEMP SPTFLTQVKE SLSSYWESAK TAAQNLYEKT YLPAVDEKLR DLYSKSTAAM STYTGIFTDQ VLSVLKGEE | |
Greater than 90% as determined by SDS-PAGE. |
344 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |