missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apolipoprotein C-II Protein
A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein C-II, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_000474.2 |
---|---|
Gene ID (Entrez) | 344 |
Name | Apolipoprotein C-II Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966829
|
enQuireBio™
QP5665-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986829
|
enQuireBio™
QP5665-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976829
|
enQuireBio™
QP5665-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15996829
|
enQuireBio™
QP5665-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15906839
|
enQuireBio™
QP5665-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15916839
|
enQuireBio™
QP5665-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000474.2 | |
Apolipoprotein C-II Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
344 | |
Research Use Only | |
APOC2 | |
Recombinant Protein | |
E. coli | |
TQQPQQDEMP SPTFLTQVKE SLSSYWESAK TAAQNLYEKT YLPAVDEKLR DLYSKSTAAM STYTGIFTDQ VLSVLKGEE | |
Greater than 90% as determined by SDS-PAGE. |