missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOH Protein
A cDNA sequence encoding the APOH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11054-100ug
Additional Details : Weight : 0.01000kg
Specifications
350 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MISPVLILFS SFLCHVAIAG RTCPKPDDLP FSTVVPLKTF YEPGEEITYS CKPGYVSRGG MRKFICPLTG LWPINTLKCT PRVCPFAGIL ENGAVRYTTF EYPNTISFSC NTGFYLNGAD SAKCTEEGKW SPELPVCAPI ICPPPSIPTF ATLRVYKPSA GNNSLYRDTA VFECLPQHAM FGNDTITCTT HGNWTKLPEC REVKCPFPSR PDNGFVNYPA KPTLYYKDKA TFGCHDGYSL DGPEEIECTK LGNWSAMPSC KASCKVPVKK ATVVYQGERV KIQEKFKNGM LHGDKVSFFC KNKEKKCSYT EDAQCIDGTI EVPKCFKEHS SLAFWKTDAS DVKPC | |
Greater than 95% as determined by SDS-PAGE. |
APOH Protein | |
Research Use Only | |
APOH | |
Human | |
Untagged | |
APOH is supplied in 16mM HEPES buffer pH 7.2, 200mM NaCl and 20% glycerol. |