missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOH Protein
A cDNA sequence encoding the APOH was constructed and used to recombinantly synthesize the protein.
£155.00 - £3635.00
Specifications
Gene ID (Entrez) | 350 |
---|---|
Name | APOH Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | APOH |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945998
|
enQuireBio™
QP11054-100UG |
100 μg |
£155.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955998
|
enQuireBio™
QP11054-1MG |
1 mg |
£835.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965998
|
enQuireBio™
QP11054-5MG |
5 mg |
£3635.00
5mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
350 | |
Research Use Only | |
APOH | |
Human | |
Untagged | |
APOH is supplied in 16mM HEPES buffer pH 7.2, 200mM NaCl and 20% glycerol. |
APOH Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MISPVLILFS SFLCHVAIAG RTCPKPDDLP FSTVVPLKTF YEPGEEITYS CKPGYVSRGG MRKFICPLTG LWPINTLKCT PRVCPFAGIL ENGAVRYTTF EYPNTISFSC NTGFYLNGAD SAKCTEEGKW SPELPVCAPI ICPPPSIPTF ATLRVYKPSA GNNSLYRDTA VFECLPQHAM FGNDTITCTT HGNWTKLPEC REVKCPFPSR PDNGFVNYPA KPTLYYKDKA TFGCHDGYSL DGPEEIECTK LGNWSAMPSC KASCKVPVKK ATVVYQGERV KIQEKFKNGM LHGDKVSFFC KNKEKKCSYT EDAQCIDGTI EVPKCFKEHS SLAFWKTDAS DVKPC | |
Greater than 95% as determined by SDS-PAGE. |