missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOB Protein
A cDNA sequence encoding the APOB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11047-10mg
Additional Details : Weight : 0.01000kg
Specifications
APOB Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
The protein was lyophilized from a 0.5 mg/ml solution of 50mM Na2CO3 pH 10 containing 50mM NaCl and 10mM sodium deoxycholate. |
10 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 95.0%. |