missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APOB Protein
A cDNA sequence encoding the APOB was constructed and used to recombinantly synthesize the protein.
£173.00 - £4955.00
Specifications
Name | APOB Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955978
|
enQuireBio™
QP11047-200UG |
200 μg |
£173.00
200µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945978
|
enQuireBio™
QP11047-1MG |
1 mg |
£660.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935978
|
enQuireBio™
QP11047-10MG |
10 mg |
£4955.00
10mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
APOB Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 95.0%. |
Research Use Only | |
Recombinant Protein | |
Human | |
The protein was lyophilized from a 0.5 mg/ml solution of 50mM Na2CO3 pH 10 containing 50mM NaCl and 10mM sodium deoxycholate. |