missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Amine oxidase [flavin-containing] B Protein
A DNA sequence encoding the Homo sapiens (Human) Amine oxidase [flavin-containing] B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6334-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000889.3 | |
Amine oxidase [flavin-containing] B Protein | |
Research Use Only | |
MAOB | |
Recombinant Protein | |
E. coli | |
SNKCDVVVVG GGISGMAAAK LLHDSGLNVV VLEARDRVGG RTYTLRNQKV KYVDLGGSYV GPTQNRILRL AKELGLETYK VNEVERLIHH VKGKSYPFRG PFPPVWNPIT YLDHNNFWRT MDDMGREIPS DAPWKAPLAE EWDNMTMKEL LDKLCWTESA KQLATLFVNL CVTAETHEVS ALWFLWYVKQ CGGTTRIIST TNGGQERKFV GGSGQVSERI MDLLGDRVKL ERPVIYIDQT RENVLVETLN HEMYEAKYVI SAIPPTLGMK IHFNPPLPMM RNQMITRVPL GSVIKCIVYY KEPFWRKKDY CGTMIIDGEE APVAYTLDDT KPEGNYAAIM GFILAHKARK LARLTKEERL KKLCELYAKV LGSLEALEPV HYEEKNWCEE QYSGGCYTTY FPPGILTQYG RVLRQPVDRI YFAGTETATH WSGYMEGAVE AGERAAREIL HAMGKIPEDE IWQSEPESVD VPAQPITTTF LERHLPSV | |
Greater than 90% as determined by SDS-PAGE. |
4129 | |
10 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |