missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Amine oxidase [flavin-containing] B Protein
A DNA sequence encoding the Homo sapiens (Human) Amine oxidase [flavin-containing] B, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_000889.3 |
---|---|
Gene ID (Entrez) | 4129 |
Name | Amine oxidase [flavin-containing] B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16072540
|
enQuireBio™
QP6334-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16092540
|
enQuireBio™
QP6334-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16082540
|
enQuireBio™
QP6334-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002550
|
enQuireBio™
QP6334-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012550
|
enQuireBio™
QP6334-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16022550
|
enQuireBio™
QP6334-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000889.3 | |
Amine oxidase [flavin-containing] B Protein | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4129 | |
Research Use Only | |
MAOB | |
Recombinant Protein | |
E. coli | |
SNKCDVVVVG GGISGMAAAK LLHDSGLNVV VLEARDRVGG RTYTLRNQKV KYVDLGGSYV GPTQNRILRL AKELGLETYK VNEVERLIHH VKGKSYPFRG PFPPVWNPIT YLDHNNFWRT MDDMGREIPS DAPWKAPLAE EWDNMTMKEL LDKLCWTESA KQLATLFVNL CVTAETHEVS ALWFLWYVKQ CGGTTRIIST TNGGQERKFV GGSGQVSERI MDLLGDRVKL ERPVIYIDQT RENVLVETLN HEMYEAKYVI SAIPPTLGMK IHFNPPLPMM RNQMITRVPL GSVIKCIVYY KEPFWRKKDY CGTMIIDGEE APVAYTLDDT KPEGNYAAIM GFILAHKARK LARLTKEERL KKLCELYAKV LGSLEALEPV HYEEKNWCEE QYSGGCYTTY FPPGILTQYG RVLRQPVDRI YFAGTETATH WSGYMEGAVE AGERAAREIL HAMGKIPEDE IWQSEPESVD VPAQPITTTF LERHLPSV | |
Greater than 90% as determined by SDS-PAGE. |