missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALK-3 / BMPR1A Protein
A DNA sequence encoding the Homo sapiens (Human) ALK-3 / BMPR1A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5725-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004320.2 | |
ALK-3 / BMPR1A Protein | |
Research Use Only | |
BMPR1A | |
Recombinant Protein | |
E. coli | |
KHYCKSISSR RRYNRDLEQD EAFIPVGESL KDLIDQSQSS GSGSGLPLLV QRTIAKQIQM VRQVGKGRYG EVWMGKWRGE KVAVKVFFTT EEASWFRETE IYQTVLMRHE NILGFIAADI KGTGSWTQLY LITDYHENGS LYDFLKCATL DTRALLKLAY SAACGLCHLH TEIYGTQGKP AIAHRDLKSK NILIKKNGSC CIADLGLAVK FNSDTNEVDV PLNTRVGTKR YMAPEVLDES LNKNHFQPYI MADIYSFGLI IWEMARRCIT GGIVEEYQLP YYNMVPSDPS YEDMREVVCV KRLRPIVSNR WNSDECLRAV LKLMSECWAH NPASRLTALR IKKTLAKMVE SQDVKI | |
Greater than 90% as determined by SDS-PAGE. |
657 | |
50 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |