missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ALK-3 / BMPR1A Protein
A DNA sequence encoding the Homo sapiens (Human) ALK-3 / BMPR1A, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_004320.2 |
---|---|
Gene ID (Entrez) | 657 |
Name | ALK-3 / BMPR1A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15927289
|
enQuireBio™
QP5725-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947289
|
enQuireBio™
QP5725-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937289
|
enQuireBio™
QP5725-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15957289
|
enQuireBio™
QP5725-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15967289
|
enQuireBio™
QP5725-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15977289
|
enQuireBio™
QP5725-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004320.2 | |
ALK-3 / BMPR1A Protein | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
657 | |
Research Use Only | |
BMPR1A | |
Recombinant Protein | |
E. coli | |
KHYCKSISSR RRYNRDLEQD EAFIPVGESL KDLIDQSQSS GSGSGLPLLV QRTIAKQIQM VRQVGKGRYG EVWMGKWRGE KVAVKVFFTT EEASWFRETE IYQTVLMRHE NILGFIAADI KGTGSWTQLY LITDYHENGS LYDFLKCATL DTRALLKLAY SAACGLCHLH TEIYGTQGKP AIAHRDLKSK NILIKKNGSC CIADLGLAVK FNSDTNEVDV PLNTRVGTKR YMAPEVLDES LNKNHFQPYI MADIYSFGLI IWEMARRCIT GGIVEEYQLP YYNMVPSDPS YEDMREVVCV KRLRPIVSNR WNSDECLRAV LKLMSECWAH NPASRLTALR IKKTLAKMVE SQDVKI | |
Greater than 90% as determined by SDS-PAGE. |