missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aldehyde oxidase Protein
A DNA sequence encoding the Homo sapiens (Human) Aldehyde oxidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5656-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001150.3 | |
Aldehyde oxidase Protein | |
Research Use Only | |
AOX1 | |
Recombinant Protein | |
E. coli | |
FGSERMMWFS PVTLKELLEF KFKYPQAPVI MGNTSVGPEV KFKGVFHPVI ISPDRIEELS VVNHAYNGLT LGAGLSLAQV KDILADVVQK LPEEKTQMYH ALLKHLGTLA GSQIRNMASL GGHIISRHPD SDLNPILAVG NCTLNLLSKE GKRQIPLNEQ FLSKCPNADL KPQEILVSVN IPYSRK | |
Greater than 90% as determined by SDS-PAGE. |
316 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |