missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aldehyde oxidase Protein
A DNA sequence encoding the Homo sapiens (Human) Aldehyde oxidase, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001150.3 |
---|---|
Gene ID (Entrez) | 316 |
Name | Aldehyde oxidase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966769
|
enQuireBio™
QP5656-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986769
|
enQuireBio™
QP5656-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976769
|
enQuireBio™
QP5656-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15996769
|
enQuireBio™
QP5656-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15906779
|
enQuireBio™
QP5656-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15916779
|
enQuireBio™
QP5656-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001150.3 | |
Aldehyde oxidase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
316 | |
Research Use Only | |
AOX1 | |
Recombinant Protein | |
E. coli | |
FGSERMMWFS PVTLKELLEF KFKYPQAPVI MGNTSVGPEV KFKGVFHPVI ISPDRIEELS VVNHAYNGLT LGAGLSLAQV KDILADVVQK LPEEKTQMYH ALLKHLGTLA GSQIRNMASL GGHIISRHPD SDLNPILAVG NCTLNLLSKE GKRQIPLNEQ FLSKCPNADL KPQEILVSVN IPYSRK | |
Greater than 90% as determined by SDS-PAGE. |