missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 78 kDa glucose-regulated Protein
A DNA sequence encoding the Homo sapiens (Human) 78 kDa glucose-regulated protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8488-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005338.1 | |
78 kDa glucose-regulated Protein | |
Research Use Only | |
HSPA5 | |
Recombinant Protein | |
E. coli | |
EDVGTVVGID LGTTYSCVGV FKNGRVEIIA NDQGNRITPS YVAFTPEGER LIGDAAKNQL TSNPENTVFD AKRLIGRTWN DPSVQQDIKF LPFKVVEKKT KPYIQVDIGG GQTKTFAPEE ISAMVLTKMK ETAEAYLGKK VTHAVVTVPA YFNDAQRQAT KDAGTIAGLN VMRIINEPTA AAIAYGLDKR EGEKNILVFD LGGGTFDVSL LTIDNGVFEV VATNGDTHLG GEDFDQRVME HFIKLYKKKT GKDVRKDNRA VQKLRREVE | |
Greater than 93.3% as determined by SDS-PAGE. |
3309 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |