missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 78 kDa glucose-regulated Protein
A DNA sequence encoding the Homo sapiens (Human) 78 kDa glucose-regulated protein, was expressed in the hosts and tags indicated.
£145.46 - £1688.38
Specifications
Accession Number | NP_005338.1 |
---|---|
Gene ID (Entrez) | 3309 |
Name | 78 kDa glucose-regulated Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094891
|
enQuireBio™
QP8488-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034901
|
enQuireBio™
QP8488-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084891
|
enQuireBio™
QP8488-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014901
|
enQuireBio™
QP8488-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024901
|
enQuireBio™
QP8488-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004901
|
enQuireBio™
QP8488-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005338.1 | |
78 kDa glucose-regulated Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3309 | |
Research Use Only | |
HSPA5 | |
Recombinant Protein | |
E. coli | |
EDVGTVVGID LGTTYSCVGV FKNGRVEIIA NDQGNRITPS YVAFTPEGER LIGDAAKNQL TSNPENTVFD AKRLIGRTWN DPSVQQDIKF LPFKVVEKKT KPYIQVDIGG GQTKTFAPEE ISAMVLTKMK ETAEAYLGKK VTHAVVTVPA YFNDAQRQAT KDAGTIAGLN VMRIINEPTA AAIAYGLDKR EGEKNILVFD LGGGTFDVSL LTIDNGVFEV VATNGDTHLG GEDFDQRVME HFIKLYKKKT GKDVRKDNRA VQKLRREVE | |
Greater than 93.3% as determined by SDS-PAGE. |