missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV HTLV-1 gp21 Protein

A cDNA sequence encoding the HTLV-1 gp21 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12354-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15980149

  • £660.00 / 500µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV HTLV-1 gp21 Protein
Research Use Only
Recombinant Protein
E. coli
60mM NaCl, 50mM Tris-HCl, pH 8.0, 0.25% Sarkosil, 50% Glycerol and 10mM Glutathione.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
GST
HTLV-1 gp21 is >95% pure as determined by 10% SDS-PAGE (coomassie staining) and HPLC-C4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers