missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV HTLV-1 gp21 Protein
A cDNA sequence encoding the HTLV-1 gp21 was constructed and used to recombinantly synthesize the protein.
£173.00 - £1320.00
Specifications
Name | HIV HTLV-1 gp21 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960149
|
enQuireBio™
QP12354-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15980149
|
enQuireBio™
QP12354-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970149
|
enQuireBio™
QP12354-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HIV HTLV-1 gp21 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
GST | |
HTLV-1 gp21 is >95% pure as determined by 10% SDS-PAGE (coomassie staining) and HPLC-C4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
60mM NaCl, 50mM Tris-HCl, pH 8.0, 0.25% Sarkosil, 50% Glycerol and 10mM Glutathione. |