missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV HTLV-1 gp21 Protein
A cDNA sequence encoding the HTLV-1 gp21 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12354-1mg
Additional Details : Weight : 0.01000kg
Specifications
HIV HTLV-1 gp21 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
60mM NaCl, 50mM Tris-HCl, pH 8.0, 0.25% Sarkosil, 50% Glycerol and 10mM Glutathione. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
GST | |
HTLV-1 gp21 is >95% pure as determined by 10% SDS-PAGE (coomassie staining) and HPLC-C4. |