missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 p24 gag Protein

A cDNA sequence encoding the HIV-1 p24 gag was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12257-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979848

  • £1320.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV-1 p24 gag Protein
Research Use Only
Recombinant Protein
E. coli
slyn tvatlycvhq rieikdtkea ldkikeeqnk skkkaqqaaa dtghssqvsq nypivqniqg qmvhqaispr tlnawvkvve ekafspevip mfsalsegat pqdlntmlnt vgghqaamqm lketineeaa ewdrvhpvha gpiapgqmre prgsdiagtt stlqeqigwm tnnppipvge iykrwiilgl nkivrmyspt sildirqgpk epfrdyvdrf yktlraeqas qevknwmtet llvqnanpdc ktilkalgpa atleemmtac qgvggpghka rvlaeamsqv tnsatimmqr gnfrnqrkiv kcfncgkegh iarncraprk kgcwkcgkeg hqmkdcterq anflgk
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
8M urea, 20mM Tris-HCl pH 8 and 10mM b-mercaptoethanol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers