missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 p24 gag Protein
A cDNA sequence encoding the HIV-1 p24 gag was constructed and used to recombinantly synthesize the protein.
£173.00 - £1320.00
Specifications
Name | HIV-1 p24 gag Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969848
|
enQuireBio™
QP12257-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989848
|
enQuireBio™
QP12257-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979848
|
enQuireBio™
QP12257-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HIV-1 p24 gag Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
8M urea, 20mM Tris-HCl pH 8 and 10mM b-mercaptoethanol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
slyn tvatlycvhq rieikdtkea ldkikeeqnk skkkaqqaaa dtghssqvsq nypivqniqg qmvhqaispr tlnawvkvve ekafspevip mfsalsegat pqdlntmlnt vgghqaamqm lketineeaa ewdrvhpvha gpiapgqmre prgsdiagtt stlqeqigwm tnnppipvge iykrwiilgl nkivrmyspt sildirqgpk epfrdyvdrf yktlraeqas qevknwmtet llvqnanpdc ktilkalgpa atleemmtac qgvggpghka rvlaeamsqv tnsatimmqr gnfrnqrkiv kcfncgkegh iarncraprk kgcwkcgkeg hqmkdcterq anflgk | |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |