missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis B HBV X Protein

A cDNA sequence encoding the HBV X was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12134-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15959478

  • £155.00 / 25µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Hepatitis B HBV X Protein
Research Use Only
X
Hepatitis B
Untagged
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 30mM acetate buffer pH4.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MAARVCCQLD PARDVLCLRP VGAESRGRPV SGPFGTLPSP SSSAVPADHG AHLSLRGLPV CAFSSAGPCA LRFTSARRME TTVNAHQVLP KVLHKRTLGL SAMSTTDLEA YFKDCLFKDW EELGEEIRLK VFVLGGCRHK LVCSPAPCNF FTSA
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers