missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBV X Protein
A cDNA sequence encoding the HBV X was constructed and used to recombinantly synthesize the protein.
£99.15 - £4075.00
Spezifikation
Name | Hepatitis B HBV X Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | X |
Product Type | Recombinant Protein |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
15969478
|
enQuireBio™
QP12134-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959478
|
enQuireBio™
QP12134-25UG |
25 μg |
£155.00
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949478
|
enQuireBio™
QP12134-1MG |
1 mg |
£4075.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Spezifikation
Hepatitis B HBV X Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAARVCCQLD PARDVLCLRP VGAESRGRPV SGPFGTLPSP SSSAVPADHG AHLSLRGLPV CAFSSAGPCA LRFTSARRME TTVNAHQVLP KVLHKRTLGL SAMSTTDLEA YFKDCLFKDW EELGEEIRLK VFVLGGCRHK LVCSPAPCNF FTSA | |
Greater than 95% as determined by SDS-PAGE. |
Research Use Only | |
X | |
Hepatitis B | |
Untagged | |
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 30mM acetate buffer pH4. |