missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBsAg adr CHO Protein
A cDNA sequence encoding the HBsAg adr CHO was constructed and used to recombinantly synthesize the protein.
£99.15 - £945.00
Specifications
Name | Hepatitis B HBsAg adr CHO Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis B |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939448
|
enQuireBio™
QP12124-20UG |
20 μg |
£99.15
20µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919448
|
enQuireBio™
QP12124-100UG |
100 μg |
£155.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929448
|
enQuireBio™
QP12124-1MG |
1 mg |
£945.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis B HBsAg adr CHO Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
Untagged | |
Sterile Filtered solution containing 20mM PB and 154mM NaCl, pH 7.1. |
Research Use Only | |
Recombinant Protein | |
Hepatitis B | |
MENTASGFLG PLLVLQAGFF LLTRILTIPQ SLDSWWTSLN FLGGAPTCPG QNSQSPTSNH SPTSCPPICP GYRWMCLRRF IIFLFILLLC LIFLLVLLDY HGMLPVCPLL PGTSTTSTGP CKTCTIPAQG TSMFPSCCCT KPSDGNCTCI PIPSSWAFAR FLWEWASVRF SWLSLLVPFV QWFVGLSPTV WLSVIWMMWY WGPSLYNILS PFLPLLPIFF CLWVYI | |
Greater than 95.0% as determined by RP-HPLC. |