missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBsAg adr CHO Protein
A cDNA sequence encoding the HBsAg adr CHO was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12124-20ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis B HBsAg adr CHO Protein | |
Research Use Only | |
Recombinant Protein | |
Hepatitis B | |
MENTASGFLG PLLVLQAGFF LLTRILTIPQ SLDSWWTSLN FLGGAPTCPG QNSQSPTSNH SPTSCPPICP GYRWMCLRRF IIFLFILLLC LIFLLVLLDY HGMLPVCPLL PGTSTTSTGP CKTCTIPAQG TSMFPSCCCT KPSDGNCTCI PIPSSWAFAR FLWEWASVRF SWLSLLVPFV QWFVGLSPTV WLSVIWMMWY WGPSLYNILS PFLPLLPIFF CLWVYI | |
Greater than 95.0% as determined by RP-HPLC. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
Untagged | |
Sterile Filtered solution containing 20mM PB and 154mM NaCl, pH 7.1. |