missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli S-ribosylhomocysteine lyase Protein
A DNA sequence encoding the Escherichia coli (strain K12) S-ribosylhomocysteine lyase, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_417172.1 |
---|---|
Gene ID (Entrez) | 947168 |
Name | S-ribosylhomocysteine lyase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16029080
|
enQuireBio™
QP7508-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049080
|
enQuireBio™
QP7508-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16039080
|
enQuireBio™
QP7508-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059080
|
enQuireBio™
QP7508-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069080
|
enQuireBio™
QP7508-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079080
|
enQuireBio™
QP7508-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_417172.1 | |
S-ribosylhomocysteine lyase Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
947168 | |
Research Use Only | |
luxS | |
Recombinant Protein | |
E. coli | |
PLLDSFTVDH TRMEAPAVRV AKTMNTPHGD AITVFDLRFC VPNKEVMPER GIHTLEHLFA GFMRNHLNGN GVEIIDISPM GCRTGFYMSL IGTPDEQRVA DAWKAAMEDV LKVQDQNQIP ELNVYQCGTY QMHSLQEAQD IARSILERDV RINSNEELAL PKEKLQELHI | |
Greater than 90% as determined by SDS-PAGE. |