missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Recombination protein RecR Protein
A DNA sequence encoding the Escherichia coli (strain K12) Recombination protein RecR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7320-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_415005.1 | |
Recombination protein RecR Protein | |
Research Use Only | |
recR | |
Recombinant Protein | |
E. coli | |
MQTSPLLTQL MEALRCLPGV GPKSAQRMAF TLLQRDRSGG MRLAQALTRA MSEIGHCADC RTFTEQEVCN ICSNPRRQEN GQICVVESPA DIYAIEQTGQ FSGRYFVLMG HLSPLDGIGP DDIGLDRLEQ RLAEEKITEV ILATNPTVEG EATANYIAEL CAQYDVEASR IAHGVPVGGE LEMVDGTTLS HSLAGRHKIR F | |
Greater than 90% as determined by SDS-PAGE. |
945100 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |