missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Recombination protein RecR Protein
A DNA sequence encoding the Escherichia coli (strain K12) Recombination protein RecR, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_415005.1 |
---|---|
Gene ID (Entrez) | 945100 |
Name | Recombination protein RecR Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028180
|
enQuireBio™
QP7320-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048180
|
enQuireBio™
QP7320-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038180
|
enQuireBio™
QP7320-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058180
|
enQuireBio™
QP7320-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068180
|
enQuireBio™
QP7320-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078180
|
enQuireBio™
QP7320-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_415005.1 | |
Recombination protein RecR Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
945100 | |
Research Use Only | |
recR | |
Recombinant Protein | |
E. coli | |
MQTSPLLTQL MEALRCLPGV GPKSAQRMAF TLLQRDRSGG MRLAQALTRA MSEIGHCADC RTFTEQEVCN ICSNPRRQEN GQICVVESPA DIYAIEQTGQ FSGRYFVLMG HLSPLDGIGP DDIGLDRLEQ RLAEEKITEV ILATNPTVEG EATANYIAEL CAQYDVEASR IAHGVPVGGE LEMVDGTTLS HSLAGRHKIR F | |
Greater than 90% as determined by SDS-PAGE. |