missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Recombination protein RecR Protein

A DNA sequence encoding the Escherichia coli (strain K12) Recombination protein RecR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7320-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048180

  • £2480.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_415005.1
Recombination protein RecR Protein
Research Use Only
recR
Recombinant Protein
E. coli
MQTSPLLTQL MEALRCLPGV GPKSAQRMAF TLLQRDRSGG MRLAQALTRA MSEIGHCADC RTFTEQEVCN ICSNPRRQEN GQICVVESPA DIYAIEQTGQ FSGRYFVLMG HLSPLDGIGP DDIGLDRLEQ RLAEEKITEV ILATNPTVEG EATANYIAEL CAQYDVEASR IAHGVPVGGE LEMVDGTTLS HSLAGRHKIR F
Greater than 90% as determined by SDS-PAGE.
945100
1 mg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers