missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Cell division protein FtsZ Protein
A DNA sequence encoding the Escherichia coli O6:H1 (strain CFT073 / ATCC 700928 / UPEC) Cell division protein FtsZ, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | WP_000462776.1 |
---|---|
Name | Cell division protein FtsZ Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | ftsZ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028210
|
enQuireBio™
QP7325-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048210
|
enQuireBio™
QP7325-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16038210
|
enQuireBio™
QP7325-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058210
|
enQuireBio™
QP7325-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068210
|
enQuireBio™
QP7325-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078210
|
enQuireBio™
QP7325-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000462776.1 | |
Research Use Only | |
ftsZ | |
Recombinant Protein | |
E. coli | |
MFEPMELTND AVIKVIGVGG GGGNAVEHMV RERIEGVEFF AVNTDAQALR KTAVGQTIQI GSGITKGLGA GANPEVGRNA ADEDRDALRA ALEGADMVFI AAGMGGGTGT GAAPVVAEVA KDLGILTVAV VTKPFNFEGK KRMAFAEQGI TELSKHVDSL ITIPNDKLLK VLGRGISLLD AFGAANDVLK GAVQGIAELI TRPGLMNVDF ADVRTVMSEM GYAMMGSGVA SGEDRAEEAA EMAISSPLLE DIDLSGARGV LVNITAGFDL RLDEFETVGN TIRAFASDNA TVVIGTSLDP DMNDELRVTV VATGIGMDKR PEITLVTNKQ VQQPVMDRYQ QHGMAPLTQE QKPVAKVVND NAPQTAKEPD YLDIPAFLRK QAD | |
Greater than 90% as determined by SDS-PAGE. |
Cell division protein FtsZ Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |