missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Cell division protein FtsZ Protein

A DNA sequence encoding the Escherichia coli O6:H1 (strain CFT073 / ATCC 700928 / UPEC) Cell division protein FtsZ, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7325-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048210

  • £2480.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_000462776.1
1 mg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Cell division protein FtsZ Protein
Research Use Only
ftsZ
Recombinant Protein
E. coli
MFEPMELTND AVIKVIGVGG GGGNAVEHMV RERIEGVEFF AVNTDAQALR KTAVGQTIQI GSGITKGLGA GANPEVGRNA ADEDRDALRA ALEGADMVFI AAGMGGGTGT GAAPVVAEVA KDLGILTVAV VTKPFNFEGK KRMAFAEQGI TELSKHVDSL ITIPNDKLLK VLGRGISLLD AFGAANDVLK GAVQGIAELI TRPGLMNVDF ADVRTVMSEM GYAMMGSGVA SGEDRAEEAA EMAISSPLLE DIDLSGARGV LVNITAGFDL RLDEFETVGN TIRAFASDNA TVVIGTSLDP DMNDELRVTV VATGIGMDKR PEITLVTNKQ VQQPVMDRYQ QHGMAPLTQE QKPVAKVVND NAPQTAKEPD YLDIPAFLRK QAD
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers