missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog Serum amyloid A Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Serum amyloid A protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6646-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024790

  • £2480.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001003050.1
Serum amyloid A Protein
Research Use Only
SAA1
Recombinant Protein
E. coli
QWYSFVSEAA QGAWDMWRAY SDMREANYKN SDKYFHARGN YDAAQRGPGG AWAAKVISDA RENSQRITDL LRFGDSGHGA EDSKADQAAN EWGRSGKDPN HFRPAGLPDK Y
Greater than 90% as determined by SDS-PAGE.
403585
1 mg
Not determined.
Full Length
Dog
His-GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers