missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Serum amyloid A Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Serum amyloid A protein, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_001003050.1 |
---|---|
Gene ID (Entrez) | 403585 |
Name | Serum amyloid A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16004790
|
enQuireBio™
QP6646-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024790
|
enQuireBio™
QP6646-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16014790
|
enQuireBio™
QP6646-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034790
|
enQuireBio™
QP6646-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044790
|
enQuireBio™
QP6646-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054790
|
enQuireBio™
QP6646-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001003050.1 | |
Serum amyloid A Protein | |
Not determined. | |
Full Length | |
Dog | |
His-GST | |
Tris-based buffer, 50% glycerol |
403585 | |
Research Use Only | |
SAA1 | |
Recombinant Protein | |
E. coli | |
QWYSFVSEAA QGAWDMWRAY SDMREANYKN SDKYFHARGN YDAAQRGPGG AWAAKVISDA RENSQRITDL LRFGDSGHGA EDSKADQAAN EWGRSGKDPN HFRPAGLPDK Y | |
Greater than 90% as determined by SDS-PAGE. |