missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog PNLIPRP1 / PLRP1 Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) PNLIPRP1 / PLRP1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6509-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16083800

  • £2480.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001003319.2
PNLIPRP1 / PLRP1 Protein
Research Use Only
PNLIPRP1
Recombinant Protein
E. coli
KEVCYEQIGC FSDAEPWAGT AIRPLKVLPW SPERIGTRFL LYTNKNPNNF QTLLPSDPST IEASNFQTDK KTRFIIHGFI DKGEENWLLD MCKNMFKVEE VNCICVDWKK GSQTSYTQAA NNVRVVGAQV AQMLSMLSAN YSYSPSQVQL IGHSLGAHVA GEAGSRTPGL GRITGLDPVE ASFQGTPEEV RLDPTDADFV DVIHTDAAPL IPFLGFGTSQ QMGHLDFFPN GGEEMPGCKK NALSQIVDLD GIWEGTRDFV ACNHLRSYKY YSESILNPDG FASYPCASYR AFESNKCFPC PDQGCPQMGH YADKFAVKTS DETQKYFLNT GDSSNFARWR YGVSITLSGK RATGQAKVAL FGSKGNTHQF NIFKGILKPG STHSNEFDAK LDVGTIEKVK FLWNNNVVNP TFPKVGAAKI TVQKGEEKTV HSFCSESTVR EDVLLTLTPC
Greater than 90% as determined by SDS-PAGE.
404010
1 mg
Not determined.
Full Length
Dog
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers