missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog PNLIPRP1 / PLRP1 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) PNLIPRP1 / PLRP1, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_001003319.2 |
---|---|
Gene ID (Entrez) | 404010 |
Name | PNLIPRP1 / PLRP1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063800
|
enQuireBio™
QP6509-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083800
|
enQuireBio™
QP6509-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073800
|
enQuireBio™
QP6509-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093800
|
enQuireBio™
QP6509-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003810
|
enQuireBio™
QP6509-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013810
|
enQuireBio™
QP6509-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spezifikation
NP_001003319.2 | |
PNLIPRP1 / PLRP1 Protein | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
404010 | |
Research Use Only | |
PNLIPRP1 | |
Recombinant Protein | |
E. coli | |
KEVCYEQIGC FSDAEPWAGT AIRPLKVLPW SPERIGTRFL LYTNKNPNNF QTLLPSDPST IEASNFQTDK KTRFIIHGFI DKGEENWLLD MCKNMFKVEE VNCICVDWKK GSQTSYTQAA NNVRVVGAQV AQMLSMLSAN YSYSPSQVQL IGHSLGAHVA GEAGSRTPGL GRITGLDPVE ASFQGTPEEV RLDPTDADFV DVIHTDAAPL IPFLGFGTSQ QMGHLDFFPN GGEEMPGCKK NALSQIVDLD GIWEGTRDFV ACNHLRSYKY YSESILNPDG FASYPCASYR AFESNKCFPC PDQGCPQMGH YADKFAVKTS DETQKYFLNT GDSSNFARWR YGVSITLSGK RATGQAKVAL FGSKGNTHQF NIFKGILKPG STHSNEFDAK LDVGTIEKVK FLWNNNVVNP TFPKVGAAKI TVQKGEEKTV HSFCSESTVR EDVLLTLTPC | |
Greater than 90% as determined by SDS-PAGE. |