missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Serotype 3 NS1 Protein

Recombinant Protein

£421.67 - £1529.90

Specifications

Name Serotype 3 NS1 Protein
Product Type Recombinant Protein
Protein Tag Native
Host Species Human
Cross Reactivity Dengue virus (Strain: Sri Lanka D3/H/IMTSSA-SRI/2000/1266)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15953998
View Documents Promotion Details
enQuireBio™
QP10480-100UG
100 μg
£421.67
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15963998
View Documents Promotion Details
enQuireBio™
QP10480-500UG
500 μg
£1529.90
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The NS1, non-structural protein, is an immunogenic protein, but poses no health risks and is therefore ideal for antibody production.
Specifications

Specifications

Serotype 3 NS1 Protein
Native
Dengue virus (Strain: Sri Lanka D3/H/IMTSSA-SRI/2000/1266)
Recombinant Protein
Human
>95% as determined by SDS-PAGE.
SDS
Product Certifications