Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 3 NS1 Protein
Recombinant Protein
Brand:  enQuireBio™ QP10480-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15953998
  • £421.00 / 100µg
Estimated Shipment: 03-06-2024
to see stock.
Description
Description
The NS1, non-structural protein, is an immunogenic protein, but poses no health risks and is therefore ideal for antibody production.
Specifications
Specifications
Serotype 3 NS1 Protein
Research Use Only
Recombinant Protein
Human
DPBS pH 7.4.
100 μg
Untested
Dengue virus (Strain: Sri Lanka D3/H/IMTSSA-SRI/2000/1266)
Native
>95% as determined by SDS-PAGE.