Learn More
enQuireBio™ Recombinant Dengue virus Dengue Virus Serotype 4 VLP Protein
Recombinant Protein
Brand: enQuireBio™ QP10484-250ug
Additional Details : Weight : 0.01000kg
Specifications
Serotype 4 VLP Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4. |
250 μg | |
Untested | |
Dengue virus | |
Native | |
>95% as determined by SDS-PAGE. |