missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue virus Dengue Virus Serotype 4 VLP Protein
Recombinant Protein
£945.00 - £1900.00
Specifications
Name | Serotype 4 VLP Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Untested |
Product Type | Recombinant Protein |
Cross Reactivity | Dengue virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934008
|
enQuireBio™
QP10484-100UG |
100 μg |
£945.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944008
|
enQuireBio™
QP10484-250UG |
250 μg |
£1900.00
250µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Serotype 4 VLP Protein | |
Untested | |
Dengue virus | |
Native | |
>95% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Human | |
10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4. |