Learn More
enQuireBio™ Recombinant Dengue virus Dengue Virus Serotype 4 VLP Protein
Recombinant Protein
Brand:  enQuireBio™ QP10484-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15934008
  • £945.00 / 100µg
Estimated Shipment: 21-06-2024
to see stock.
Specifications
Serotype 4 VLP Protein
Research Use Only
Recombinant Protein
Human
10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4.
100 μg
Untested
Dengue virus
Native
>95% as determined by SDS-PAGE.