missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue NS1 ST2 Protein
A cDNA sequence encoding the Dengue NS1 ST2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5285.00
Specifications
Name | Dengue NS1 ST2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Dengue Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987898
|
enQuireBio™
QP11645-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15967898
|
enQuireBio™
QP11645-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977898
|
enQuireBio™
QP11645-1MG |
1 mg |
£5285.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Dengue NS1 ST2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
His | |
Protein is >95% pure as determined by 12.5% SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
Dengue NS1 ST2 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, Aprotinin and Pepstatin A. |