missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue NS1 ST2 Protein

A cDNA sequence encoding the Dengue NS1 ST2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11645-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967898

  • £155.00 / 10µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Dengue NS1 ST2 Protein
Research Use Only
Recombinant Protein
Baculovirus
Dengue NS1 ST2 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, Aprotinin and Pepstatin A.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
His
Protein is >95% pure as determined by 12.5% SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers