missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cotton Rat IL-13 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£326.13
Specifications
Molecular Weight (g/mol) | 13.1 kDa |
---|---|
Name | IL-13 |
Quantity | 25μg |
Source | Chinese Hamster Ovary cell line,CHO-derived cotton rat IL-13 protein Met1-His133,with a C-terminal 5-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18389457
|
R&D Systems™
6940-CL-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£326.13
25µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18167680
|
R&D Systems™
6940-CL-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£326.13
25µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Cotton Rat IL-13 Protein is derived from CHO. The Recombinant Cotton Rat IL-13 Protein has been validated for the following applications: Bioactivity.Specifications
13.1 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
IL-13 | |
Chinese Hamster Ovary cell line,CHO-derived cotton rat IL-13 protein Met1-His133,with a C-terminal 5-His tag | |
Monomer | |
Recombinant |