missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cotton Rat IL-13 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 6940-CL-025/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18167680

  • £326.13 / 25µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Cotton Rat IL-13 Protein is derived from CHO. The Recombinant Cotton Rat IL-13 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
IL-13
Chinese Hamster Ovary cell line,CHO-derived cotton rat IL-13 protein Met1-His133,with a C-terminal 5-His tag
Monomer
Recombinant
13.1 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers